Learn More
4-Methoxy-2-nitroaniline, 97%, Thermo Scientific Chemicals
£19.80 - £110.00
Chemical Identifiers
CAS | 96-96-8 |
---|---|
Molecular Formula | C7H8N2O3 |
Molecular Weight (g/mol) | 168.15 |
MDL Number | MFCD00007152 |
InChI Key | QFMJFXFXQAFGBO-UHFFFAOYSA-N |
Synonym | 2-nitro-4-methoxyaniline, 4-amino-3-nitroanisole, fast bordeaux gp, 2-nitro-p-anisidine, azobase nas, devol bordeaux b, bordeaux gp base, bordeaux gp salt, fast bordeaux gp base, fast bordeaux gp salt |
PubChem CID | 66793 |
ChEBI | CHEBI:48973 |
IUPAC Name | 4-methoxy-2-nitroaniline |
SMILES | COC1=CC(=C(C=C1)N)[N+](=O)[O-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10061270
|
Thermo Scientific Acros
125960250 |
25 g | Glass bottle |
£19.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10615752
|
Thermo Scientific Acros
125961000 |
100 g | Glass bottle |
£29.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10153270
|
Thermo Scientific Acros
125965000 |
500 g | Glass bottle |
£110.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
96-96-8 | |
168.15 | |
QFMJFXFXQAFGBO-UHFFFAOYSA-N | |
66793 | |
4-methoxy-2-nitroaniline |
C7H8N2O3 | |
MFCD00007152 | |
2-nitro-4-methoxyaniline, 4-amino-3-nitroanisole, fast bordeaux gp, 2-nitro-p-anisidine, azobase nas, devol bordeaux b, bordeaux gp base, bordeaux gp salt, fast bordeaux gp base, fast bordeaux gp salt | |
CHEBI:48973 | |
COC1=CC(=C(C=C1)N)[N+](=O)[O-] |
Specifications
96-96-8 | |
100.0 | |
96% min. (HPLC) | |
CH3OC6H3(NO2)NH2 | |
13, 521 | |
Solubility in water: slightly soluble | |
COC1=CC(=C(C=C1)N)[N+](=O)[O-] | |
168.15 | |
CHEBI:48973 | |
97% |
96.0 | |
123.0°C to 126.0°C | |
C7H8N2O3 | |
MFCD00007152 | |
2-nitro-4-methoxyaniline, 4-amino-3-nitroanisole, fast bordeaux gp, 2-nitro-p-anisidine, azobase nas, devol bordeaux b, bordeaux gp base, bordeaux gp salt, fast bordeaux gp base, fast bordeaux gp salt | |
QFMJFXFXQAFGBO-UHFFFAOYSA-N | |
4-methoxy-2-nitroaniline | |
66793 | |
168.15 | |
4-Methoxy-2-nitroaniline, 98% |
Safety and Handling
GHS H Statement:
Fatal in contact with skin.
Fatal if swallowed.
Fatal if inhaled.
Harmful to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-547-2
missing translation for 'rtecsNumber' : BY4415000
missing translation for 'tsca' : TSCA
- 4-Methoxy-2-nitroaniline
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- H300-Fatal if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P361-Take off immediately all contaminated clothing.
RUO – Research Use Only