missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-(trifluoromethoxy)benzoic acid, 97%, Thermo Scientific™

Product Code. 15456367
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 647855-22-9
Molecular Formula C9H7F3O4
Molecular Weight (g/mol) 236.146
MDL Number MFCD18398451
InChI Key PHSHCIBRNMLSBJ-UHFFFAOYSA-N
Synonym 4-methoxy-3-trifluoromethoxy benzoic acid, benzoic acid, 4-methoxy-3-trifluoromethoxy, 4-methoxy-3-trifluoromethoxy-benzoic acid
PubChem CID 66523504
IUPAC Name 4-methoxy-3-(trifluoromethoxy)benzoic acid
SMILES COC1=C(C=C(C=C1)C(=O)O)OC(F)(F)F

Specifications

Quantity 5 g
Formula Weight 236.14
Percent Purity 97%
Chemical Name or Material 4-Methoxy-3-(trifluoromethoxy)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'