missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-(trifluoromethyl)benzaldehyde, 98%, Thermo Scientific™

Product Code. 11382119
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11382119

Brand: Thermo Scientific Alfa Aesar H26128.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Slightly soluble in water.

Notes
Air Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50823-87-5
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.148
MDL Number MFCD01091012
InChI Key UMZQFCFPWAFMRL-UHFFFAOYSA-N
Synonym 4-methoxy-3-trifluoromethyl benzaldehyde, 3-trifluoromethyl-4-methoxybenzaldehyde, pubchem4247, acmc-1aqw0, 3-trifluoromethyl-p-anisaldehyde, benzaldehyde, 4-methoxy-3-trifluoromethyl
PubChem CID 2775291
IUPAC Name 4-methoxy-3-(trifluoromethyl)benzaldehyde
SMILES COC1=C(C=C(C=C1)C=O)C(F)(F)F

Specifications

Melting Point 38°C to 40°C
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 204.15
Percent Purity 98%
Chemical Name or Material 4-Methoxy-3-(trifluoromethyl)benzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.