missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-(trifluoromethyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 15449995
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149507-36-8
Molecular Formula C8H8BF3O3
Molecular Weight (g/mol) 219.954
MDL Number MFCD07363790
InChI Key BUSMBMGODABSIN-UHFFFAOYSA-N
Synonym 4-methoxy-3-trifluoromethyl phenyl boronic acid, 4-methoxy-3-trifluoromethylphenylboronic acid, 3-trifluoromethyl-4-methoxy-phenylboronic acid, 4-methoxy-3-trifluoromethylphenyl boronic acid, 4-methoxy-3-trifluoromethyl benzeneboronic acid, 4-methoxy-3-trifluoromethyl phenylboronic acid, boronic acid, 4-methoxy-3-trifluoromethyl phenyl, acmc-1c5ik
PubChem CID 17750052
IUPAC Name [4-methoxy-3-(trifluoromethyl)phenyl]boronic acid
SMILES B(C1=CC(=C(C=C1)OC)C(F)(F)F)(O)O

Specifications

Melting Point 198°C to 202°C
Quantity 250 mg
Formula Weight 219.95
Percent Purity 98%
Chemical Name or Material 4-Methoxy-3-(trifluoromethyl)benzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'