missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 11409757
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 213598-09-5
Molecular Formula C9H7F3O3
Molecular Weight (g/mol) 220.15
MDL Number MFCD01091014
InChI Key VNWUPESTVBTYJM-UHFFFAOYSA-N
Synonym 4-methoxy-3-trifluoromethyl benzoic acid, 4-methoxy-3-trifluoromethyl-benzoic acid, acmc-1cr7h, 3-trifluoromethyl-4-methoxybenzoic acid, benzoic acid, 4-methoxy-3-trifluoromethyl, 4-methyloxy-3-trifluoromethyl benzoic acid
PubChem CID 2775295
IUPAC Name 4-methoxy-3-(trifluoromethyl)benzoic acid
SMILES COC1=CC=C(C=C1C(F)(F)F)C(O)=O

Specifications

Melting Point 169°C to 171°C
Quantity 5 g
Beilstein 8619237
Formula Weight 220.15
Percent Purity 97%
Chemical Name or Material 4-Methoxy-3-(trifluoromethyl)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'