Learn More
4'-Methoxyacetophenone, 98%, Thermo Scientific Chemicals
£32.00 - £82.40
Chemical Identifiers
CAS | 100-06-1 |
---|---|
Molecular Formula | C9H10O2 |
Molecular Weight (g/mol) | 150.18 |
MDL Number | MFCD00008745 |
InChI Key | NTPLXRHDUXRPNE-UHFFFAOYSA-N |
Synonym | 4'-methoxyacetophenone, 4-acetylanisole, 4-methoxyacetophenone, 1-4-methoxyphenyl ethanone, p-methoxyacetophenone, acetanisole, novatone, linarodin, vananote, ethanone, 1-4-methoxyphenyl |
PubChem CID | 7476 |
ChEBI | CHEBI:86567 |
SMILES | COC1=CC=C(C=C1)C(C)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10226010
|
Thermo Scientific Acros
152362500 |
250 g | Glass bottle |
£32.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10091710
|
Thermo Scientific Acros
152360010 |
1 kg | Glass bottle |
£82.40
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100-06-1 | |
150.18 | |
NTPLXRHDUXRPNE-UHFFFAOYSA-N | |
7476 | |
COC1=CC=C(C=C1)C(C)=O |
C9H10O2 | |
MFCD00008745 | |
4'-methoxyacetophenone, 4-acetylanisole, 4-methoxyacetophenone, 1-4-methoxyphenyl ethanone, p-methoxyacetophenone, acetanisole, novatone, linarodin, vananote, ethanone, 1-4-methoxyphenyl | |
CHEBI:86567 |
Specifications
100-06-1 | |
100.0 | |
White | |
138°C | |
97.5% min. (GC) | |
CH3OC6H4COCH3 | |
08, 87 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether | |
COC1=CC=C(C=C1)C(C)=O | |
150.18 | |
CHEBI:86567 | |
98% | |
4'-Methoxyacetophenone, 98% |
97.5 | |
36.0°C to 39.0°C | |
260.0°C | |
Authentic | |
C9H10O2 | |
MFCD00008745 | |
4'-methoxyacetophenone, 4-acetylanisole, 4-methoxyacetophenone, 1-4-methoxyphenyl ethanone, p-methoxyacetophenone, acetanisole, novatone, linarodin, vananote, ethanone, 1-4-methoxyphenyl | |
NTPLXRHDUXRPNE-UHFFFAOYSA-N | |
1-(4-methoxyphenyl)ethanone | |
7476 | |
150.18 | |
Crystalline Powder or Crystals |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 202-815-9
missing translation for 'rtecsNumber' : AM9240000
missing translation for 'tsca' : TSCA
- 4'-Methoxyacetophenone
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only