missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxybenzenediazonium tetrafluoroborate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
4-Methoxybenzenediazonium tetrafluoroborate has been used in the preparation of azo coupled cyclic β-enaminones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methoxybenzenediazonium tetrafluoroborate has been used in the preparation of azo coupled cyclic β-enaminones.
Solubility
Decompose in water
Notes
Store at -20°C. Light sensitive.
4-Methoxybenzenediazonium tetrafluoroborate has been used in the preparation of azo coupled cyclic β-enaminones.
Solubility
Decompose in water
Notes
Store at -20°C. Light sensitive.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 142°C to 144°C |
Quantity | 1 g |
UN Number | UN3261 |
Solubility Information | Decompose in water |
IUPAC Name | 4-methoxybenzenediazonium;tetrafluoroborate |
Formula Weight | 221.95 |
Percent Purity | 98% |
Chemical Name or Material | 4-Methoxybenzenediazonium tetrafluoroborate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'