missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxybenzyl carbazate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
It finds its application in the derived azide in N-protection and as a starting material for the azide used to introduce the pMZ-amino protection which is removable under mild hydrolytic conditions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds its application in the derived azide in N-protection and as a starting material for the azide used to introduce the pMZ-amino protection which is removable under mild hydrolytic conditions.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It finds its application in the derived azide in N-protection and as a starting material for the azide used to introduce the pMZ-amino protection which is removable under mild hydrolytic conditions.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
Melting Point | 75°C to 77°C |
Quantity | 1 g |
Beilstein | 2052164 |
Solubility Information | Insoluble in water. |
Formula Weight | 196.21 |
Percent Purity | 97% |
Chemical Name or Material | 4-Methoxybenzyl carbazate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'