missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxybenzyl isothiocyanate, 94%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
4-Methoxybenzyl isothiocyanate is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methoxybenzyl isothiocyanate is used as pharmaceutical intermediates.
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Incompatible with bases, alcohols and amines. Store under dry inert gas. Protect from humidity and water. Store away from moisture.
4-Methoxybenzyl isothiocyanate is used as pharmaceutical intermediates.
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Incompatible with bases, alcohols and amines. Store under dry inert gas. Protect from humidity and water. Store away from moisture.
Specifications
Specifications
Density | 1.15 |
Boiling Point | 132°C to 133°C (0.7 mmHg) |
Odor | Irritating |
Refractive Index | 1.5975 |
Quantity | 5 g |
UN Number | UN2922 |
Beilstein | 2089549 |
Sensitivity | Moisture sensitive |
Solubility Information | Hydrolyzes in water. |
Formula Weight | 179.24 |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction