Learn More
4-(Methoxycarbonyl)phenyl isothiocyanate, 98+%, Thermo Scientific Chemicals
£48.70 - £166.00
Chemical Identifiers
CAS | 3662-78-0 |
---|---|
Molecular Formula | C9H7NO2S |
Molecular Weight (g/mol) | 193.22 |
MDL Number | MFCD00041099 |
InChI Key | WIZODHILGBTPPA-UHFFFAOYSA-N |
Synonym | 4-methoxycarbonylphenyl isothiocyanate, 4-methoxycarbonylphenylisothiocyanate, 4-isothiocyanatobenzoic acid methyl ester, benzoic acid, 4-isothiocyanato-, methyl ester, acmc-20amez, #, 4-methoxycarbonyl phenylisothiocyanate, 4-methoxycarbonyl phenyl isothiocyanate |
PubChem CID | 138002 |
IUPAC Name | methyl 4-isothiocyanatobenzoate |
SMILES | COC(=O)C1=CC=C(C=C1)N=C=S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11388445
|
Thermo Scientific Alfa Aesar
L10474.03 |
1 g |
£48.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11398445
|
Thermo Scientific Alfa Aesar
L10474.06 |
5 g |
£166.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3662-78-0 | |
193.22 | |
WIZODHILGBTPPA-UHFFFAOYSA-N | |
138002 | |
COC(=O)C1=CC=C(C=C1)N=C=S |
C9H7NO2S | |
MFCD00041099 | |
4-methoxycarbonylphenyl isothiocyanate, 4-methoxycarbonylphenylisothiocyanate, 4-isothiocyanatobenzoic acid methyl ester, benzoic acid, 4-isothiocyanato-, methyl ester, acmc-20amez, #, 4-methoxycarbonyl phenylisothiocyanate, 4-methoxycarbonyl phenyl isothiocyanate | |
methyl 4-isothiocyanatobenzoate |
Specifications
3662-78-0 | |
Irritating | |
MFCD00041099 | |
2100504 | |
4-methoxycarbonylphenyl isothiocyanate, 4-methoxycarbonylphenylisothiocyanate, 4-isothiocyanatobenzoic acid methyl ester, benzoic acid, 4-isothiocyanato-, methyl ester, acmc-20amez, #, 4-methoxycarbonyl phenylisothiocyanate, 4-methoxycarbonyl phenyl isothiocyanate | |
COC(=O)C1=CC=C(C=C1)N=C=S | |
193.22 | |
193.23 | |
4-(Methoxycarbonyl)phenyl isothiocyanate |
46°C to 50°C | |
C9H7NO2S | |
UN2923 | |
Moisture sensitive | |
WIZODHILGBTPPA-UHFFFAOYSA-N | |
methyl 4-isothiocyanatobenzoate | |
138002 | |
≥98% |
Safety and Handling
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312-H314-H331-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only