missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenol, 98+%

Product Code. 11462617
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11462617

Brand: Thermo Scientific Alfa Aesar A14845.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 150-76-5 | C7H8O2 | 124.14 g/mol

4-Methoxyphenol is an active ingredient and used in dermatology. It is employed as a pharmaceutical drug in skin depigmentation. It is used as polymerization inhibitors. For example, in the radical polymerization of acryaltes and styrene monomers. It is also used as an intermediate in the preparationagrochemicals, liquid crystals. It acts as a stabilizer for the formulation of inks, toners and adhesives. It is mainly used as an additive for textile and leather industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxyphenol is an active ingredient and used in dermatology. It is employed as a pharmaceutical drug in skin depigmentation. It is used as polymerization inhibitors. For example, in the radical polymerization of acryaltes and styrene monomers. It is also used as an intermediate in the preparationagrochemicals, liquid crystals. It acts as a stabilizer for the formulation of inks, toners and adhesives. It is mainly used as an additive for textile and leather industries.

Solubility
Soluble in acetone, ethyl acetate, ethanol, ether, benzene and carbon tetrachloride.

Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed. Incompatible with halogens and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-76-5
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00002332
InChI Key NWVVVBRKAWDGAB-UHFFFAOYSA-N
Synonym mequinol, 4-hydroxyanisole, p-hydroxyanisole, p-methoxyphenol, hydroquinone monomethyl ether, phenol, 4-methoxy, leucobasal, mehq, leucodine b, mechinolum
PubChem CID 9015
ChEBI CHEBI:69441
IUPAC Name 4-methoxyphenol
SMILES COC1=CC=C(O)C=C1

Specifications

Melting Point 55°C to 58°C
Density 1.55
Boiling Point 243°C to 246°C
Flash Point 132°C (269°F)
Quantity 1000 g
Beilstein 507924
Solubility Information Soluble in acetone,ethyl acetate,ethanol,ether,benzene and carbon tetrachloride.
Formula Weight 124.14
Percent Purity ≥98%
Chemical Name or Material 4-Methoxyphenol
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.