missing translation for 'onlineSavingsMsg'
Learn More
4-Methoxyphenol, 99%, Thermo Scientific Chemicals Product Code.: 10605562

4-Methoxyphenol, 99%, Thermo Scientific Chemicals

Product Code. 10605562
1 kg
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
25 kg
This item is not returnable. View return policy

Product Code. 10605562

Brand: Thermo Scientific Acros 126000010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-76-5, 123-31-9
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00002332
InChI Key NWVVVBRKAWDGAB-UHFFFAOYSA-N
Synonym mequinol, 4-hydroxyanisole, p-hydroxyanisole, p-methoxyphenol, hydroquinone monomethyl ether, phenol, 4-methoxy, leucobasal, mehq, leucodine b, mechinolum
PubChem CID 9015
ChEBI CHEBI:69441
IUPAC Name 4-methoxyphenol
SMILES COC1=CC=C(O)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 56.0°C to 57.0°C
Color Colorless to Yellow
Boiling Point 243.0°C
Flash Point 133°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Quantity 1 kg
Beilstein 06,843
Solubility Information Solubility in water: 40g/L (25°C). Other solubilities: readily soluble in ethyl acetate, 455g/100g ethyl alcohol - 426g/100g acetone (25°C), 70g/100g benzene - 3g/100g carbon tetrachloride, (25°C)
Viscosity 8.5 mPa.s (65°C)
Formula Weight 124.14
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material 4-Methoxyphenol, Stabilized
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methoxyphenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Methoxyphenol, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.