missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenoxyacetic acid, 98%, Thermo Scientific Chemicals

Product Code. 11407057
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1877-75-4
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00014360
InChI Key BHFSBJHPPFJCOS-UHFFFAOYSA-N
Synonym 4-methoxyphenoxyacetic acid, 2-4-methoxyphenoxy acetic acid, 4-methoxyphenoxy acetic acid, acetic acid, 4-methoxyphenoxy, p-methoxyphenoxyacetic acid, acetic acid, p-methoxyphenoxy, acetic acid, 2-4-methoxyphenoxy, p-methoxyphenoxy acetic acid, akos bb/0049, rarechem al bo 0321
PubChem CID 74649
IUPAC Name 2-(4-methoxyphenoxy)acetic acid
SMILES COC1=CC=C(OCC(O)=O)C=C1

Specifications

Melting Point 109°C to 113°C
Quantity 250 g
Beilstein 1874579
Formula Weight 182.18
Percent Purity 98%
Chemical Name or Material 4-Methoxyphenoxyacetic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'