missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl beta-D-glucopyranoside, 98%, Thermo Scientific™

Product Code. 15497769
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6032-32-2
Molecular Formula C13H18O7
Molecular Weight (g/mol) 286.28
MDL Number MFCD06797143
InChI Key SIXFVXJMCGPTRB-UJPOAAIJSA-N
Synonym methylarbutin, 4-methoxyphenyl beta-d-glucopyranoside, p-methoxyphenyl b-d-glucoside, unii-f9mw5fu0xh, 4-methoxyphenyl b-d-glucopyranoside, f9mw5fu0xh, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-4-methoxyphenoxy oxane-3,4,5-triol, methylarbutoside, 4-methoxyphenylglucoside, p-methoxyphenyl beta-d-glucopyranoside
PubChem CID 80131
ChEBI CHEBI:81207
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxane-3,4,5-triol
SMILES COC1=CC=C(C=C1)OC2C(C(C(C(O2)CO)O)O)O

Specifications

Melting Point 175°C to 179°C
Color White
Quantity 1 g
Solubility Information Soluble in DMSO or water
Optical Rotation −64.5° (c=1 in Water)
Formula Weight 286.28
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Methoxyphenyl beta-D-glucopyranoside
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'