Learn More
4-Methoxyphenylacetyl chloride, 98%, Thermo Scientific Chemicals
£78.70 - £261.00
Chemical Identifiers
CAS | 4693-91-8 |
---|---|
Molecular Formula | C9H9ClO2 |
Molecular Weight (g/mol) | 184.619 |
MDL Number | MFCD00075474 |
InChI Key | CXJOONIFSVSFAD-UHFFFAOYSA-N |
Synonym | 4-methoxyphenylacetyl chloride, 2-4-methoxyphenyl acetyl chloride, benzenacetyl chloride, 4-methoxy, benzeneacetyl chloride, 4-methoxy, 4-methoxyphenylacetylchloride, p-methoxyphenyl acetyl chloride, p-methoxyphenylacetyl chloride, acmc-1aso1, 4-methoxyphenylacetylchlorid, 4-methoxybenzeneacetyl chloride |
PubChem CID | 2734688 |
IUPAC Name | 2-(4-methoxyphenyl)acetyl chloride |
SMILES | COC1=CC=C(C=C1)CC(=O)Cl |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4693-91-8 | |
184.619 | |
CXJOONIFSVSFAD-UHFFFAOYSA-N | |
2734688 | |
COC1=CC=C(C=C1)CC(=O)Cl |
C9H9ClO2 | |
MFCD00075474 | |
4-methoxyphenylacetyl chloride, 2-4-methoxyphenyl acetyl chloride, benzenacetyl chloride, 4-methoxy, benzeneacetyl chloride, 4-methoxy, 4-methoxyphenylacetylchloride, p-methoxyphenyl acetyl chloride, p-methoxyphenylacetyl chloride, acmc-1aso1, 4-methoxyphenylacetylchlorid, 4-methoxybenzeneacetyl chloride | |
2-(4-methoxyphenyl)acetyl chloride |
Specifications
4693-91-8 | |
143°C (10 mmHg) | |
C9H9ClO2 | |
MFCD00075474 | |
908061 | |
4-methoxyphenylacetyl chloride, 2-4-methoxyphenyl acetyl chloride, benzenacetyl chloride, 4-methoxy, benzeneacetyl chloride, 4-methoxy, 4-methoxyphenylacetylchloride, p-methoxyphenyl acetyl chloride, p-methoxyphenylacetyl chloride, acmc-1aso1, 4-methoxyphenylacetylchlorid, 4-methoxybenzeneacetyl chloride | |
COC1=CC=C(C=C1)CC(=O)Cl | |
184.619 | |
184.62 | |
4-Methoxyphenylacetyl chloride |
1.208 | |
>110°C (230°F) | |
1.54 | |
UN3265 | |
Moisture sensitive | |
CXJOONIFSVSFAD-UHFFFAOYSA-N | |
2-(4-methoxyphenyl)acetyl chloride | |
2734688 | |
98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only