Learn More
4-Methoxyphenylacetylene, 98%, Thermo Scientific Chemicals
£37.10 - £116.00
Chemical Identifiers
CAS | 768-60-5 |
---|---|
Molecular Formula | C9H8O |
Molecular Weight (g/mol) | 132.162 |
MDL Number | MFCD00168815 |
InChI Key | KBIAVTUACPKPFJ-UHFFFAOYSA-N |
Synonym | 4-ethynylanisole, 4-methoxyphenylacetylene, 4'-methoxyphenyl acetylene, 1-ethynyl-4-methoxy-benzene, 1-eth-1-ynyl-4-methoxybenzene, benzene, 1-ethynyl-4-methoxy, 4-methoxyphenyl acetylene, 4-ethynyl-1-methoxybenzene, 4'-methoxyphenylacetylene, p-ethynylanisole |
PubChem CID | 251020 |
IUPAC Name | 1-ethynyl-4-methoxybenzene |
SMILES | COC1=CC=C(C=C1)C#C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11356779
|
Thermo Scientific Alfa Aesar
H26283.03 |
1 g |
£37.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11366779
|
Thermo Scientific Alfa Aesar
H26283.06 |
5 g |
£116.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Ethynylanisole was used in the synthesis of photo luminescent 1,2-dihydrophosphinines via a [4 + 2] cycloaddition. Along with an arylboronic acid and sodium azide in a copper-catalyzed, three-component synthesis of trisubstituted 1,2,4-triazoles. In a study of a gold (III)-catalyzed hydroamination of alkynes leading to N-vinylindoles3.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Ethynylanisole was used in the synthesis of photo luminescent 1,2-dihydrophosphinines via a [4 + 2] cycloaddition. Along with an arylboronic acid and sodium azide in a copper-catalyzed, three-component synthesis of trisubstituted 1,2,4-triazoles. In a study of a gold (III)-catalyzed hydroamination of alkynes leading to N-vinylindoles3.
Solubility
Insoluble in water. Soluble in chloroform, acetone, dichloromethane, and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
768-60-5 | |
132.162 | |
KBIAVTUACPKPFJ-UHFFFAOYSA-N | |
251020 | |
COC1=CC=C(C=C1)C#C |
C9H8O | |
MFCD00168815 | |
4-ethynylanisole, 4-methoxyphenylacetylene, 4'-methoxyphenyl acetylene, 1-ethynyl-4-methoxy-benzene, 1-eth-1-ynyl-4-methoxybenzene, benzene, 1-ethynyl-4-methoxy, 4-methoxyphenyl acetylene, 4-ethynyl-1-methoxybenzene, 4'-methoxyphenylacetylene, p-ethynylanisole | |
1-ethynyl-4-methoxybenzene |
Specifications
768-60-5 | |
1.019 | |
82°C (180°F) | |
1.563 | |
4-ethynylanisole, 4-methoxyphenylacetylene, 4'-methoxyphenyl acetylene, 1-ethynyl-4-methoxy-benzene, 1-eth-1-ynyl-4-methoxybenzene, benzene, 1-ethynyl-4-methoxy, 4-methoxyphenyl acetylene, 4-ethynyl-1-methoxybenzene, 4'-methoxyphenylacetylene, p-ethynylanisole | |
KBIAVTUACPKPFJ-UHFFFAOYSA-N | |
1-ethynyl-4-methoxybenzene | |
251020 | |
99% |
27°C to 29°C | |
87°C to 91°C (11 mmHg) | |
C9H8O | |
MFCD00168815 | |
Insoluble in water. Soluble in chloroform,acetone,dichloromethane,and methanol. | |
COC1=CC=C(C=C1)C#C | |
132.162 | |
132.16 | |
4-Methoxyphenylacetylene |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335-H500
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only