missing translation for 'onlineSavingsMsg'
Learn More
4-Methoxythiophenol, 97%, Thermo Scientific Chemicals Product Code.: 11408626

4-Methoxythiophenol, 97%, Thermo Scientific Chemicals

Product Code. 11408626
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11408626

Brand: Thermo Scientific Alfa Aesar A16009.06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Methoxythiophenol is used in preparing samples for self-assembled monolayers (SAM?s) characterization and to study SAM’s effect on both n- and p-channel organic thin film transistors. It is also used in cesium fluoride-Celite catalyzed preparation of thioethers and thioesters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxythiophenol is used in preparing samples for self-assembled monolayers (SAM′s) characterization and to study SAM’s effect on both n- and p-channel organic thin film transistors. It is also used in cesium fluoride-Celite catalyzed preparation of thioethers and thioesters.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 696-63-9
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
MDL Number MFCD00004849
InChI Key NIFAOMSJMGEFTQ-UHFFFAOYSA-N
Synonym 4-methoxythiophenol, p-methoxybenzenethiol, benzenethiol, 4-methoxy, 4-methoxybenzene-1-thiol, benzenethiol, p-methoxy, 4-mercaptoanisole, 4-methoxy thiophenol, 4-methoxy-benzenethiol, para-methoxybenzenethiol, 4-mercaptoanisol
PubChem CID 12765
IUPAC Name 4-methoxybenzenethiol
SMILES COC1=CC=C(C=C1)S

Specifications

Density 1.14
Boiling Point 100°C to 103°C (13 mmHg)
Flash Point 96°C (204°F)
Odor Unpleasant
Refractive Index 1.5831
Quantity 5 g
Beilstein 1446910
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix.
Formula Weight 140.21
Percent Purity 97%
Chemical Name or Material 4-Methoxythiophenol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Methoxythiophenol, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.