missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxytrityl chloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
It is used as selective protecting reagent that is used for primary hydroxyl groups.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as selective protecting reagent that is used for primary hydroxyl groups.
Solubility
Soluble in toluene.
Notes
It is hygroscopic. Incompatible with oxidizing agents.
It is used as selective protecting reagent that is used for primary hydroxyl groups.
Solubility
Soluble in toluene.
Notes
It is hygroscopic. Incompatible with oxidizing agents.
Specifications
Melting Point | 121°C to 125°C |
Quantity | 10 g |
Beilstein | 798425 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in toluene. |
Formula Weight | 308.81 |
Percent Purity | 97% |
Chemical Name or Material | 4-Methoxytrityl chloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'