missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,2-cyclohexene oxide, cis + trans, 97%

Product Code. 11432230
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11432230

Brand: Thermo Scientific Alfa Aesar H29100.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 36099-51-1 | C7H12O | 112.172 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36099-51-1
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD09742280
InChI Key ULPDSNLBZMHGPI-UHFFFAOYSA-N
Synonym 3-methyl-7-oxabicyclo 4.1.0 heptane, 7-oxabicyclo 4.1.0 heptane, 3-methyl, 4-methyl-1,2-cyclohexene oxide, 4-methyl-1,2-cyclohexene oxide, cis + trans, 4-methyl-7-oxabicyclo 4.1.0 heptane, 7-oxabicyclo 4.1.0 heptane,3-methyl, 3-methyl-7-oxabicyclo 4.1.0 heptane #, 4-methyl-1,2-cyclohexene oxide, cis+trans
PubChem CID 535184
IUPAC Name 4-methyl-7-oxabicyclo[4.1.0]heptane
SMILES CC1CCC2C(C1)O2

Specifications

Refractive Index 1.447
Quantity 5 g
Formula Weight 112.17
Percent Purity 97%
Chemical Name or Material 4-Methyl-1,2-cyclohexene oxide, Cis + Trans

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.