missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,3-dioxane, 99%, Thermo Scientific Chemicals

Product Code. 10642793
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-97-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00006569
InChI Key INCCMBMMWVKEGJ-UHFFFAOYSA-N
Synonym 4-methyldioxane, 4-methyl-m-dioxane, 1,3-dioxane, 4-methyl, m-dioxane, 4-methyl, 1,3-butanediol formal, 4-methyl-1,3 dioxane, acmc-2099cl, 5-19-01-00054 beilstein handbook reference, 4-methyl-1,3-dioxan-2-yl, m-dioxane, 4-methyl-8ci
PubChem CID 14269
IUPAC Name 4-methyl-1,3-dioxane
SMILES CC1CCOCO1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -45°C to 44°C
Color Colorless
Density 0.9700g/mL
Boiling Point 114°C
Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4140 to 1.416
Quantity 5 g
Specific Gravity 0.97
Formula Weight 102.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Methyl-1, 3-dioxane, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.