missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1-pentenylboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 15423747
Click to view available options
Quantity:
50 mg
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 214907-33-2
Molecular Formula C6H13BO2
Molecular Weight (g/mol) 127.98
MDL Number MFCD01074680
InChI Key OLTGERMANPOCTC-UHFFFAOYSA-N
Synonym 4-methyl-1-pentenylboronic acid, 4-methylpent-1-en-1-yl boronic acid, trans-4-methyl-1-pentenylboronic acid, e-4-methylpent-1-enyl boronic acid, e-4-methylpent-1-en-1-yl boronic acid, 1e-4-methylpent-1-en-1-ylboronic acid
PubChem CID 6081507
IUPAC Name [(E)-4-methylpent-1-enyl]boronic acid
SMILES CC(C)CC=CB(O)O

Specifications

Melting Point 99°C to 102°C
Quantity 250 mg
Formula Weight 127.98
Percent Purity 97%
Chemical Name or Material 4-Methyl-1-pentenylboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'