Learn More
4-Methyl-2-pentanol, 99+%, Thermo Scientific Chemicals
£20.60 - £158.00
Chemical Identifiers
CAS | 108-11-2 |
---|---|
Molecular Formula | C6H14O |
Molecular Weight (g/mol) | 102.18 |
MDL Number | MFCD00004550 |
InChI Key | WVYWICLMDOOCFB-UHFFFAOYSA-N |
Synonym | 4-methyl-2-pentanol, 2-pentanol, 4-methyl, isobutylmethylcarbinol, 2-methyl-4-pentanol, methyl amyl alcohol, methyl isobutyl carbinol, mibc, 1,3-dimethylbutanol, 4-methylpentanol-2, isobutylmethylmethanol |
PubChem CID | 7910 |
IUPAC Name | 4-methylpentan-2-ol |
SMILES | CC(C)CC(C)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10685192
|
Thermo Scientific Acros
149380050 |
5 mL | Glass bottle |
£20.70
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10725041
|
Thermo Scientific Acros
149385000 |
500 mL | Glass bottle |
£20.60
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10143290
|
Thermo Scientific Acros
149380010 |
1 L | Glass bottle |
£29.20
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10214720
|
Thermo Scientific Acros
149380025 |
2.5 L | Glass bottle |
£56.20
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10155303
|
Thermo Scientific Acros
149380100 |
10 L | Plastic drum |
£158.00
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
108-11-2 | |
102.18 | |
WVYWICLMDOOCFB-UHFFFAOYSA-N | |
7910 | |
CC(C)CC(C)O |
C6H14O | |
MFCD00004550 | |
4-methyl-2-pentanol, 2-pentanol, 4-methyl, isobutylmethylcarbinol, 2-methyl-4-pentanol, methyl amyl alcohol, methyl isobutyl carbinol, mibc, 1,3-dimethylbutanol, 4-methylpentanol-2, isobutylmethylmethanol | |
4-methylpentan-2-ol |
Specifications
108-11-2 | |
100.0 | |
Colorless | |
132.0°C | |
Authentic | |
C6H14O | |
1.4100 to 1.4120 | |
01, 410 | |
4-methyl-2-pentanol, 2-pentanol, 4-methyl, isobutylmethylcarbinol, 2-methyl-4-pentanol, methyl amyl alcohol, methyl isobutyl carbinol, mibc, 1,3-dimethylbutanol, 4-methylpentanol-2, isobutylmethylmethanol | |
WVYWICLMDOOCFB-UHFFFAOYSA-N | |
4-methylpentan-2-ol | |
7910 | |
99+% | |
4-Methyl-2-pentanol |
99.0 | |
-90.0°C | |
0.8000g/mL | |
41°C | |
99+% | |
(CH3)2CHCH2CH(OH)CH3 | |
MFCD00004550 | |
0.8 | |
Solubility in water: 20g/L. Other solubilities: misscible with most common organic solvents | |
CC(C)CC(C)O | |
102.18 | |
102.18 | |
Liquid |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-551-7
RUO – Research Use Only