missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-2-pentanone, ACS Reagent, ≥98.5%, Honeywell™ Riedel-de Haën™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Specifications
Melting Point | -80°C |
Density | 0.801g/mL at 25°C |
Boiling Point | 118°C |
Flash Point | 14°C (57.2°F) |
Infrared Spectrum | Conforms to structure |
Packaging | Stainless Steel Can |
Linear Formula | (CH3)2CHCH2COCH3 |
Refractive Index | n20/D 1.395 |
Quantity | 20 L |
UN Number | UN1245 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction