missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
Density | 0.801g/mL at 25°C |
Flash Point | 14°C (57.2°F) |
Linear Formula | (CH3)2CHCH2COCH3 |
Quantity | 1 L |
UN Number | UN1245 |
Beilstein | 605399 |
Vapor Density | 3.5 (vs air) |
Formula Weight | 100.16g/mol |
Percent Purity | ≥99% |
Chemical Name or Material | 4-Methyl-2-pentanone |
Safety and Handling


Product Identifier
- 4-Methyl-2-pentanone
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : 4-methylpentan-2-one
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-Methyl-2-pentanone, Honeywell >
Spot an opportunity for improvement?Share a Content Correction