missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-2-pentanone, Honeywell
Click to view available options
Quantity:
1 L
2.5 L
20 kg
Spezifikation
Spezifikation
Density | 0.801g/mL at 25°C |
Flash Point | 14°C (57.2°F) |
Linear Formula | (CH3)2CHCH2COCH3 |
Quantity | 20 kg |
UN Number | UN1245 |
Beilstein | 605399 |
Vapor Density | 3.5 (vs air) |
Formula Weight | 100.16g/mol |
Percent Purity | ≥99% |
Chemical Name or Material | 4-Methyl-2-pentanone |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'