Learn More
4-Methyl-2-pentenal, 95%, Thermo Scientific Chemicals
£90.50 - £329.00
Chemical Identifiers
CAS | 5362-56-1 |
---|---|
Molecular Formula | C6H10O |
Molecular Weight (g/mol) | 98.14 |
MDL Number | MFCD00800501 |
InChI Key | RIWPMNBTULNXOH-ONEGZZNKSA-N |
Synonym | 4-methyl-2-pentenal, unii-sxg1guk91a, e-4-methyl-2-pentenal, sxg1guk91a, fema no. 3510, 2-pentenal, 4-methyl, 4-methylpent-2-enal, 4-methyl-2-penten-1-al, 2-pentenal, 4-methyl-, 2e, 2e-4-methylpent-2-enal |
PubChem CID | 5916154 |
IUPAC Name | (E)-4-methylpent-2-enal |
SMILES | CC(C)C=CC=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10165510
|
Thermo Scientific Acros
298350250 |
25 mL | Glass bottle |
£90.50
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10005513
|
Thermo Scientific Acros
298351000 |
100 mL | Glass bottle |
£329.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5362-56-1 | |
98.14 | |
RIWPMNBTULNXOH-ONEGZZNKSA-N | |
5916154 | |
CC(C)C=CC=O |
C6H10O | |
MFCD00800501 | |
4-methyl-2-pentenal, unii-sxg1guk91a, e-4-methyl-2-pentenal, sxg1guk91a, fema no. 3510, 2-pentenal, 4-methyl, 4-methylpent-2-enal, 4-methyl-2-penten-1-al, 2-pentenal, 4-methyl-, 2e, 2e-4-methylpent-2-enal | |
(E)-4-methylpent-2-enal |
Specifications
15mg KOH/g max. | |
94.0 | |
Colorless to Yellow | |
70.0°C (75.0 mmHg) | |
Authentic | |
C6H10O | |
1.4360 to 1.4440 | |
0.844 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol,propylene glycol,essential,oils | |
CC(C)C=CC=O | |
98.14 | |
98.14 | |
Liquid |
5362-56-1 | |
100.0 | |
0.8440g/mL | |
32°C | |
94% min. (GC) | |
CH3CH(CH3)CH=CHCHO | |
MFCD00800501 | |
4-methyl-2-pentenal, unii-sxg1guk91a, e-4-methyl-2-pentenal, sxg1guk91a, fema no. 3510, 2-pentenal, 4-methyl, 4-methylpent-2-enal, 4-methyl-2-penten-1-al, 2-pentenal, 4-methyl-, 2e, 2e-4-methylpent-2-enal | |
RIWPMNBTULNXOH-ONEGZZNKSA-N | |
(E)-4-methylpent-2-enal | |
5916154 | |
95% | |
4-Methyl-2-pentenal |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 226-348-5
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only