missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-2-phenyl-1,3-thiazole-5-carboxylic acid, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 219.26 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | 4-Methyl-2-phenyl-1, 3-thiazole-5-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'