missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-Methyl-2-phenyl-5-pyrimidinyl)methanol, 95%, Thermo Scientific™
£26.55 - £229.00
Chemical Identifiers
CAS | 342405-27-0 |
---|---|
Molecular Formula | C12H12N2O |
Molecular Weight (g/mol) | 200.241 |
MDL Number | MFCD02682074 |
InChI Key | LXJHDOHHZSZIMJ-UHFFFAOYSA-N |
Synonym | 4-methyl-2-phenylpyrimidin-5-yl methanol, 4-methyl-2-phenyl-5-pyrimidinyl methanol, 5-pyrimidinemethanol,4-methyl-2-phenyl, 4-methyl-2-phenylpyrimidin-5-yl methan-1-ol |
PubChem CID | 2776512 |
IUPAC Name | (4-methyl-2-phenylpyrimidin-5-yl)methanol |
SMILES | CC1=NC(=NC=C1CO)C2=CC=CC=C2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10437232
|
Thermo Scientific Maybridge
CC20209CB |
250 mg |
£26.55
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10437422
|
Thermo Scientific Maybridge
CC20209DA |
1 g |
£69.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10220275
|
Thermo Scientific Maybridge
CC20209DE |
5 g |
£229.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
342405-27-0 | |
200.241 | |
LXJHDOHHZSZIMJ-UHFFFAOYSA-N | |
2776512 | |
CC1=NC(=NC=C1CO)C2=CC=CC=C2 |
C12H12N2O | |
MFCD02682074 | |
4-methyl-2-phenylpyrimidin-5-yl methanol, 4-methyl-2-phenyl-5-pyrimidinyl methanol, 5-pyrimidinemethanol,4-methyl-2-phenyl, 4-methyl-2-phenylpyrimidin-5-yl methan-1-ol | |
(4-methyl-2-phenylpyrimidin-5-yl)methanol |
Specifications
342405-27-0 | |
100.0 | |
Amber Glass Bottle | |
MFCD02682074 | |
LXJHDOHHZSZIMJ-UHFFFAOYSA-N | |
(4-methyl-2-phenylpyrimidin-5-yl)methanol | |
2776512 | |
95% |
94.5 | |
95% | |
C12H12N2O | |
4-methyl-2-phenylpyrimidin-5-yl methanol, 4-methyl-2-phenyl-5-pyrimidinyl methanol, 5-pyrimidinemethanol,4-methyl-2-phenyl, 4-methyl-2-phenylpyrimidin-5-yl methan-1-ol | |
CC1=NC(=NC=C1CO)C2=CC=CC=C2 | |
200.241 | |
200.24 | |
(4-Methyl-2-phenyl-5-pyrimidinyl)methanol |