missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-phenylthiazole-5-carboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 15400778
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33763-20-1
Molecular Formula C11H8NO2S
Molecular Weight (g/mol) 218.25
MDL Number MFCD00141955
InChI Key CRSMRBYEBHOYRM-UHFFFAOYSA-M
Synonym 4-methyl-2-phenylthiazole-5-carboxylic acid, 5-thiazolecarboxylic acid, 4-methyl-2-phenyl, 4-methyl-2-phenyl-thiazole-5-carboxylic acid, 4-methyl-2-phenyl-5-thiazolecarboxylic acid, 2-phenyl-4-methylthiazole-5-carboxylic acid, maybridge1_008941, 5-thiazolecarboxylicacid, 4-methyl-2-phenyl, 4-methyl-2-phenyl-1,3-thiazol-5-carboxylic acid
PubChem CID 720957
IUPAC Name 4-methyl-2-phenyl-1,3-thiazole-5-carboxylic acid
SMILES CC1=C(SC(=N1)C1=CC=CC=C1)C([O-])=O

Specifications

Melting Point 202°C to 206°C
Quantity 5 g
Formula Weight 219.26
Percent Purity 97%
Chemical Name or Material 4-Methyl-2-phenylthiazole-5-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'