missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-nitrobenzeneboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 11303946
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80500-27-2
Molecular Formula C7H8BNO4
Molecular Weight (g/mol) 180.954
MDL Number MFCD00191550
InChI Key OASVXBRTNVFKFS-UHFFFAOYSA-N
Synonym 4-methyl-3-nitrophenyl boronic acid, 4-methyl-3-nitrobenzeneboronic acid, 3-nitro-p-tolylboronic acid, 4-methyl-3-nitrophenyl boranediol, 4-methyl-3-nitro-phenyl boronic acid, 3-nitro-4-methylphenylboronic acid, 2-nitrotoluene-4-boronic acid, boronic acid, 4-methyl-3-nitrophenyl, 3-nitro-4-methylbenzeneboronic acid
PubChem CID 2773515
IUPAC Name (4-methyl-3-nitrophenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)C)[N+](=O)[O-])(O)O

Specifications

Melting Point ∼250°C
Quantity 5 g
Beilstein 3279018
Formula Weight 180.96
Percent Purity 98%
Chemical Name or Material 4-Methyl-3-nitrobenzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'