Learn More
4-Methyl-3-nitrobenzoic acid, 99%, Thermo Scientific Chemicals
£22.70 - £245.00
Chemical Identifiers
CAS | 96-98-0 |
---|---|
Molecular Formula | C8H7NO4 |
Molecular Weight (g/mol) | 181.15 |
MDL Number | MFCD00007174 |
InChI Key | BBEWSMNRCUXQRF-UHFFFAOYSA-N |
PubChem CID | 7319 |
SMILES | CC1=C(C=C(C=C1)C(=O)O)[N+](=O)[O-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10091930
|
Thermo Scientific Acros
165990250 |
25 g | Glass bottle |
£22.70
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10001940
|
Thermo Scientific Acros
165991000 |
100 g | Glass bottle |
£58.10
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10051910
|
Thermo Scientific Acros
165995000 |
500 g | Glass bottle |
£245.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
96-98-0 | |
181.15 | |
BBEWSMNRCUXQRF-UHFFFAOYSA-N | |
CC1=C(C=C(C=C1)C(=O)O)[N+](=O)[O-] |
C8H7NO4 | |
MFCD00007174 | |
7319 |
Specifications
96-98-0 | |
100.0 | |
White to Yellow | |
99% | |
CH3C6H3(NO2)CO2H | |
09, 502 | |
Solubility in water: slightly soluble. Other solubilities: soluble in alcohols | |
CC1=C(C=C(C=C1)C(=O)O)[N+](=O)[O-] | |
7319 | |
99% | |
4-Methyl-3-nitrobenzoic acid, 99% |
98.5 | |
187.0°C to 190.0°C | |
Authentic | |
C8H7NO4 | |
MFCD00007174 | |
4-methyl-3-nitrobenzoic acid, 3-nitro-4-methylbenzoic acid, 3-nitro-p-toluic acid, benzoic acid, 4-methyl-3-nitro, 3-nitro-para-toluic acid, m-nitro-p-toluic acid, 4-methyl-3-nitro-benzoic acid, p-toluic acid, 3-nitro, 3-nitro-4-methyl benzoic acid, benzoic acid,4-methyl-3-nitro | |
BBEWSMNRCUXQRF-UHFFFAOYSA-N | |
181.15 | |
181.15 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 202-549-3
missing translation for 'tsca' : TSCA
RUO – Research Use Only