missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-nitrothiobenzamide, 97%, Thermo Scientific™

Product Code. 15420918
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1233513-42-2
Molecular Formula C8H8N2O2S
Molecular Weight (g/mol) 196.22
MDL Number MFCD09025715
InChI Key PGQMEJDSPAENNP-UHFFFAOYSA-N
Synonym 4-methyl-3-nitrothiobenzamide
PubChem CID 45040726
IUPAC Name 4-methyl-3-nitrobenzenecarbothioamide
SMILES CC1=C(C=C(C=C1)C(N)=S)[N+]([O-])=O

Specifications

Quantity 1 g
Formula Weight 196.22
Percent Purity 97%
Chemical Name or Material 4-Methyl-3-nitrothiobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'