missing translation for 'onlineSavingsMsg'
Learn More
(4-Methyl-6,7-dihydro-4H-thieno[3,2-c]pyran-4-yl)acetic acid, 97%, Thermo Scientific™ Product Code.: 10786725

(4-Methyl-6,7-dihydro-4H-thieno[3,2-c]pyran-4-yl)acetic acid, 97%, Thermo Scientific™

Product Code. 10786725
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10786725

Brand: Thermo Scientific Maybridge CC61801CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 57153-43-2
Molecular Formula C10H12O3S
Molecular Weight (g/mol) 212.263
MDL Number MFCD01850960
InChI Key MBZWBAMMUGJINB-UHFFFAOYSA-N
Synonym 4-methyl-6,7-dihydro-4h-thieno 3,2-c pyran-4-yl acetic acid, 4-methyl-6h,7h-thieno 3,2-c pyran-4-yl acetic acid, 4h-thieno 3,2-c pyran-4-aceticacid, 6,7-dihydro-4-methyl, 2-4-methyl-6,7-dihydro-4h-thieno 3,2-c pyran-4-yl acetic acid, 2-4-methyl-6,7-dihydro-4h-thiopheno 3,2-c pyran-4-yl acetic acid
PubChem CID 12225898
IUPAC Name 2-(4-methyl-6,7-dihydrothieno[3,2-c]pyran-4-yl)acetic acid
SMILES CC1(C2=C(CCO1)SC=C2)CC(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 212.27
Percent Purity 97%
Chemical Name or Material (4-Methyl-6, 7-dihydro-4H-thieno[3, 2-c]pyran-4-yl)acetic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(4-Methyl-6,7-dihydro-4H-thieno[3,2-c]pyran-4-yl)acetic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.