missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-pyrido[3,2-b][1,4, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 276.14 |
Percent Purity | 97% |
Chemical Name or Material | 4-Methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'