missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-N-phenylbenzamide, 97%, Thermo Scientific™

Product Code. 15453618
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6833-18-7
Molecular Formula C14H13NO
Molecular Weight (g/mol) 211.264
MDL Number MFCD00017213
InChI Key PQEOPHYIUYAVDQ-UHFFFAOYSA-N
PubChem CID 730569
IUPAC Name 4-methyl-N-phenylbenzamide
SMILES CC1=CC=C(C=C1)C(=O)NC2=CC=CC=C2

Specifications

Quantity 250 mg
Synonym 4-methylbenzanilide, benzamide, 4-methyl-n-phenyl, p-methyl benzanilide, benzamide,4-methyl-n-phenyl, cbmicro_015159, n-phenyl-4-methylbenzamide, 4-methylphenyl-n-benzamide
Formula Weight 211.26
Percent Purity 97%
Chemical Name or Material 4-Methyl-N-phenylbenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'