missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl(thiobenzamide), 97%, Thermo Scientific Chemicals

Product Code. 11489836
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2362-62-1
Molecular Formula C8H9NS
Molecular Weight (g/mol) 151.227
MDL Number MFCD00173750
InChI Key QXYZSNGZMDVLKN-UHFFFAOYSA-N
Synonym 4-methylbenzothioamide, 4-methyl thiobenzamide, 4-methylthiobenzamide, 4-methylbenzene-1-carbothioamide, 4-methyl-thiobenzamide, 4-methylbenzenethioamide, benzenecarbothioamide, 4-methyl, amino 4-methylphenyl methane-1-thione, 4-pmtb, p-methylthiobenzoylamide
PubChem CID 737209
IUPAC Name 4-methylbenzenecarbothioamide
SMILES CC1=CC=C(C=C1)C(=S)N

Specifications

Melting Point 166°C to 169°C
Quantity 5 g
Formula Weight 151.23
Percent Purity 97%
Chemical Name or Material 4-Methyl(thiobenzamide)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'