Learn More
4'-Methylacetophenone, 95%, Thermo Scientific Chemicals
£17.40 - £60.70
Chemical Identifiers
CAS | 122-00-9 |
---|---|
Molecular Formula | C9H10O |
Molecular Weight (g/mol) | 134.18 |
MDL Number | MFCD00008751 |
InChI Key | GNKZMNRKLCTJAY-UHFFFAOYSA-N |
Synonym | 4'-methylacetophenone, p-methylacetophenone, 1-p-tolyl ethanone, 1-p-tolylethanone, melilotal, 4-methylacetophenone, 4-acetyltoluene, 1-4-methylphenyl ethanone, methyl p-tolyl ketone, p-acetotoluene |
PubChem CID | 8500 |
IUPAC Name | 1-(4-methylphenyl)ethanone |
SMILES | CC(=O)C1=CC=C(C)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10215320
|
Thermo Scientific Acros
126180050 |
5 g | Glass bottle |
£17.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10636902
|
Thermo Scientific Acros
126182500 |
250 g | Glass bottle |
£25.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10417710
|
Thermo Scientific Acros
126180010 |
1 kg | Glass bottle |
£60.70
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
122-00-9 | |
134.18 | |
GNKZMNRKLCTJAY-UHFFFAOYSA-N | |
8500 | |
CC(=O)C1=CC=C(C)C=C1 |
C9H10O | |
MFCD00008751 | |
4'-methylacetophenone, p-methylacetophenone, 1-p-tolyl ethanone, 1-p-tolylethanone, melilotal, 4-methylacetophenone, 4-acetyltoluene, 1-4-methylphenyl ethanone, methyl p-tolyl ketone, p-acetotoluene | |
1-(4-methylphenyl)ethanone |
Specifications
122-00-9 | |
100.0 | |
Colorless to Yellow | |
226.0°C | |
Authentic | |
C9H10O | |
1.5320 to 1.5350 | |
07, 307 | |
4'-methylacetophenone, p-methylacetophenone, 1-p-tolyl ethanone, 1-p-tolylethanone, melilotal, 4-methylacetophenone, 4-acetyltoluene, 1-4-methylphenyl ethanone, methyl p-tolyl ketone, p-acetotoluene | |
GNKZMNRKLCTJAY-UHFFFAOYSA-N | |
1-(4-methylphenyl)ethanone | |
8500 | |
95% | |
4'-Methylacetophenone, 95% |
94.0 | |
-19.0°C | |
1.0050g/mL | |
92°C | |
94% min. (GC) | |
CH3C6H4COCH3 | |
MFCD00008751 | |
1.005 | |
Solubility in water: 0.37g/L (15°C) | |
CC(=O)C1=CC=C(C)C=C1 | |
134.18 | |
134.18 | |
Liquid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 204-514-8
missing translation for 'rtecsNumber' : AM9463000
missing translation for 'tsca' : TSCA
- 4'-Methylacetophenone
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only