missing translation for 'onlineSavingsMsg'
Learn More

4-Methylamino-1-propionylpiperidine, 97%, Thermo Scientific™

Product Code. 11469630
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 902836-34-4
Molecular Formula C9H18N2O
Molecular Weight (g/mol) 170.26
MDL Number MFCD08061056
InChI Key RRUSDXAHDXKSFA-UHFFFAOYSA-N
Synonym 4-methylamino-1-propionylpiperidine, 1-4-methylamino piperidin-1-yl propan-1-one, n-methyl-1-propionyl-4-piperidinamine, 1-4-methylamino piperidyl propan-1-one, 1-4-methylamino-piperidin-1-yl-propan-1-one
PubChem CID 24208856
IUPAC Name 1-[4-(methylamino)piperidin-1-yl]propan-1-one
SMILES CCC(=O)N1CCC(CC1)NC

Specifications

Quantity 1 g
Formula Weight 170.26
Percent Purity 97%
Chemical Name or Material 4-Methylamino-1-propionylpiperidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'