Learn More
4-(Methylamino)phenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
£172.00 - £632.00
Chemical Identifiers
CAS | 845870-55-5 |
---|---|
Molecular Formula | C13H20BNO2 |
Molecular Weight (g/mol) | 233.12 |
MDL Number | MFCD06795668 |
InChI Key | WWGNYCWKFZIQQS-UHFFFAOYSA-N |
Synonym | n-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-n-methylamino phenylboronic acid, pinacol ester, 4-n-methylamino phenylboronic acid pinacol ester, methyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-amine, 4-n-methylamino phenylboronic acid,pinacol ester, 4-methylamino phenylboronic acid pinacol ester, 4-methylamino phenyl boronic acid pinacol ester, n-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, benzenamine, n-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl |
PubChem CID | 11957892 |
IUPAC Name | N-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline |
SMILES | CNC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11976111
|
Thermo Scientific Acros
445150010 |
1 g |
£172.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11986111
|
Thermo Scientific Acros
445150050 |
5 g |
£632.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
845870-55-5 | |
233.12 | |
WWGNYCWKFZIQQS-UHFFFAOYSA-N | |
11957892 | |
CNC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1 |
C13H20BNO2 | |
MFCD06795668 | |
n-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-n-methylamino phenylboronic acid, pinacol ester, 4-n-methylamino phenylboronic acid pinacol ester, methyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-amine, 4-n-methylamino phenylboronic acid,pinacol ester, 4-methylamino phenylboronic acid pinacol ester, 4-methylamino phenyl boronic acid pinacol ester, n-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, benzenamine, n-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
N-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline |
Specifications
845870-55-5 | |
96% min. (GC) | |
C13H20BNO2 | |
n-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-n-methylamino phenylboronic acid, pinacol ester, 4-n-methylamino phenylboronic acid pinacol ester, methyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-amine, 4-n-methylamino phenylboronic acid,pinacol ester, 4-methylamino phenylboronic acid pinacol ester, 4-methylamino phenyl boronic acid pinacol ester, n-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, benzenamine, n-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
CNC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1 | |
233.12 | |
233.12 | |
4-(Methylamino)phenylboronic acid pinacol ester |
65.0°C to 66.0°C | |
Glass bottle | |
MFCD06795668 | |
WWGNYCWKFZIQQS-UHFFFAOYSA-N | |
N-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline | |
11957892 | |
97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
- 4-(Methylamino)phenylboronic acid pinacol ester
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only