missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylbenzeneboronic acid neopentyl glycol ester, 99%, Thermo Scientific Chemicals
£21.80 - £225.00
Chemical Identifiers
CAS | 380481-66-3 |
---|---|
Molecular Formula | C12H17BO2 |
Molecular Weight (g/mol) | 204.076 |
MDL Number | MFCD03788728 |
InChI Key | ZSPBWRPQSPRISS-UHFFFAOYSA-N |
Synonym | 5,5-dimethyl-2-p-tolyl-1,3,2-dioxaborinane, 4-methylbenzeneboronic acid neopentyl glycol ester, 5,5-dimethyl-2-4-methylphenyl-1,3,2-dioxaborinane, amtb973, 2-p-tolyl-5,5-dimethyl-1,3,2-dioxaborinane, 5,5-dimethyl-2-p-tolyl-1,3,2-dioxaborine, 2-4-methylphenyl-5,5-dimethyl-1,3,2-dioxaborinane |
PubChem CID | 23005406 |
IUPAC Name | 5,5-dimethyl-2-(4-methylphenyl)-1,3,2-dioxaborinane |
SMILES | B1(OCC(CO1)(C)C)C2=CC=C(C=C2)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11305335
|
Thermo Scientific Alfa Aesar
L19563.03 |
1 g |
£21.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11315335
|
Thermo Scientific Alfa Aesar
L19563.06 |
5 g |
£70.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11325335
|
Thermo Scientific Alfa Aesar
L19563.14 |
25 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
380481-66-3 | |
204.076 | |
ZSPBWRPQSPRISS-UHFFFAOYSA-N | |
23005406 | |
B1(OCC(CO1)(C)C)C2=CC=C(C=C2)C |
C12H17BO2 | |
MFCD03788728 | |
5,5-dimethyl-2-p-tolyl-1,3,2-dioxaborinane, 4-methylbenzeneboronic acid neopentyl glycol ester, 5,5-dimethyl-2-4-methylphenyl-1,3,2-dioxaborinane, amtb973, 2-p-tolyl-5,5-dimethyl-1,3,2-dioxaborinane, 5,5-dimethyl-2-p-tolyl-1,3,2-dioxaborine, 2-4-methylphenyl-5,5-dimethyl-1,3,2-dioxaborinane | |
5,5-dimethyl-2-(4-methylphenyl)-1,3,2-dioxaborinane |
Specifications
380481-66-3 | |
C12H17BO2 | |
5,5-dimethyl-2-p-tolyl-1,3,2-dioxaborinane, 4-methylbenzeneboronic acid neopentyl glycol ester, 5,5-dimethyl-2-4-methylphenyl-1,3,2-dioxaborinane, amtb973, 2-p-tolyl-5,5-dimethyl-1,3,2-dioxaborinane, 5,5-dimethyl-2-p-tolyl-1,3,2-dioxaborine, 2-4-methylphenyl-5,5-dimethyl-1,3,2-dioxaborinane | |
B1(OCC(CO1)(C)C)C2=CC=C(C=C2)C | |
204.076 | |
204.08 | |
4-Methylbenzeneboronic acid neopentyl glycol ester |
93°C to 95°C | |
MFCD03788728 | |
ZSPBWRPQSPRISS-UHFFFAOYSA-N | |
5,5-dimethyl-2-(4-methylphenyl)-1,3,2-dioxaborinane | |
23005406 | |
99% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only