missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzotrifluoride, 98%

Product Code. 11367506
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11367506

Brand: Thermo Scientific Alfa Aesar L11043.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6140-17-6 | C8H7F3 | 160.139 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6140-17-6
Molecular Formula C8H7F3
Molecular Weight (g/mol) 160.139
MDL Number MFCD00075476
InChI Key LRLRAYMYEXQKID-UHFFFAOYSA-N
Synonym 4-methylbenzotrifluoride, 1-methyl-4-trifluoromethyl benzene, 4-trifluoromethyl toluene, p-methylbenzotrifluoride, alpha,alpha,alpha-trifluoro-p-xylene, benzene, 1-methyl-4-trifluoromethyl, 4-methyl-1-trifluoromethyl benzene, 1-methyl-4-trifluormethyl benzol, pubchem1659, p-trifluoromethyltoluene
PubChem CID 80230
IUPAC Name 1-methyl-4-(trifluoromethyl)benzene
SMILES CC1=CC=C(C=C1)C(F)(F)F

Specifications

Density 1.144
Boiling Point 129°C
Flash Point 27°C (80°F)
Refractive Index 1.426
Quantity 1 g
UN Number UN1993
Beilstein 1863596
Formula Weight 160.14
Percent Purity 98%
Chemical Name or Material 4-Methylbenzotrifluoride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.