Learn More
4-Methylbenzyl isothiocyanate, 96%, Thermo Scientific Chemicals
£53.40 - £165.00
Chemical Identifiers
CAS | 3694-46-0 |
---|---|
Molecular Formula | C9H9NS |
Molecular Weight (g/mol) | 163.238 |
MDL Number | MFCD00041118 |
InChI Key | OAXIUBJXQISJEV-UHFFFAOYSA-N |
Synonym | 4-methylbenzyl isothiocyanate, 1-isothiocyanatomethyl-4-methylbenzene, p-methylbenzyl isothiocyanate, benzene, 1-isothiocyanatomethyl-4-methyl, isothiocyanic acid, p-methylbenzyl ester, benzene, 1-isothiocyanatomethyl-4-methyl-9ci, acmc-1cobu, 4-methylbenzylisothiocyanate, wln: scn1r d1, 4-12-00-02578 beilstein handbook reference |
PubChem CID | 19409 |
IUPAC Name | 1-(isothiocyanatomethyl)-4-methylbenzene |
SMILES | CC1=CC=C(C=C1)CN=C=S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11387576
|
Thermo Scientific Alfa Aesar
L11135.04 |
2 g |
£53.40
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11397576
|
Thermo Scientific Alfa Aesar
L11135.09 |
10 g |
£165.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3694-46-0 | |
163.238 | |
OAXIUBJXQISJEV-UHFFFAOYSA-N | |
19409 | |
CC1=CC=C(C=C1)CN=C=S |
C9H9NS | |
MFCD00041118 | |
4-methylbenzyl isothiocyanate, 1-isothiocyanatomethyl-4-methylbenzene, p-methylbenzyl isothiocyanate, benzene, 1-isothiocyanatomethyl-4-methyl, isothiocyanic acid, p-methylbenzyl ester, benzene, 1-isothiocyanatomethyl-4-methyl-9ci, acmc-1cobu, 4-methylbenzylisothiocyanate, wln: scn1r d1, 4-12-00-02578 beilstein handbook reference | |
1-(isothiocyanatomethyl)-4-methylbenzene |
Specifications
3694-46-0 | |
97°C (1 mmHg) | |
C9H9NS | |
MFCD00041118 | |
2082478 | |
4-methylbenzyl isothiocyanate, 1-isothiocyanatomethyl-4-methylbenzene, p-methylbenzyl isothiocyanate, benzene, 1-isothiocyanatomethyl-4-methyl, isothiocyanic acid, p-methylbenzyl ester, benzene, 1-isothiocyanatomethyl-4-methyl-9ci, acmc-1cobu, 4-methylbenzylisothiocyanate, wln: scn1r d1, 4-12-00-02578 beilstein handbook reference | |
CC1=CC=C(C=C1)CN=C=S | |
163.238 | |
163.24 | |
4-Methylbenzyl isothiocyanate |
1.08 | |
Pungent | |
1.589 | |
UN2922 | |
Moisture sensitive | |
OAXIUBJXQISJEV-UHFFFAOYSA-N | |
1-(isothiocyanatomethyl)-4-methylbenzene | |
19409 | |
96% |
Safety and Handling
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'rtecsNumber' : NX8860000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only