Learn More
4-Methylcinnamic acid, 99%, predominantly trans, Thermo Scientific™
£11.30 - £23.00
Chemical Identifiers
CAS | 1866-39-3 |
---|---|
MDL Number | MFCD00002697 |
InChI Key | RURHILYUWQEGOS-VOTSOKGWSA-N |
Synonym | 4-methylcinnamic acid, 3-p-tolyl acrylic acid, e-3-p-tolyl acrylic acid, p-methylcinnamic acid, p-methyl cinnamic acid, 2e-3-4-methylphenyl acrylic acid, 2e-3-4-methylphenyl prop-2-enoic acid, 4-methyl-cinnamic acid, e-3-4-methylphenyl-2-propenoic acid, 3-4-methylphenyl-2-propenoic acid |
PubChem CID | 731767 |
IUPAC Name | (E)-3-(4-methylphenyl)prop-2-enoic acid |
SMILES | CC1=CC=C(C=C1)C=CC(=O)O |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1866-39-3 | |
RURHILYUWQEGOS-VOTSOKGWSA-N | |
731767 | |
CC1=CC=C(C=C1)C=CC(=O)O |
MFCD00002697 | |
4-methylcinnamic acid, 3-p-tolyl acrylic acid, e-3-p-tolyl acrylic acid, p-methylcinnamic acid, p-methyl cinnamic acid, 2e-3-4-methylphenyl acrylic acid, 2e-3-4-methylphenyl prop-2-enoic acid, 4-methyl-cinnamic acid, e-3-4-methylphenyl-2-propenoic acid, 3-4-methylphenyl-2-propenoic acid | |
(E)-3-(4-methylphenyl)prop-2-enoic acid |
Specifications
1866-39-3 | |
White | |
C10H10O2 | |
MFCD00002697 | |
4-methylcinnamic acid, 3-p-tolyl acrylic acid, e-3-p-tolyl acrylic acid, p-methylcinnamic acid, p-methyl cinnamic acid, 2e-3-4-methylphenyl acrylic acid, 2e-3-4-methylphenyl prop-2-enoic acid, 4-methyl-cinnamic acid, e-3-4-methylphenyl-2-propenoic acid, 3-4-methylphenyl-2-propenoic acid | |
CC1=CC=C(C=C1)C=CC(=O)O | |
162.19 | |
162.19 | |
Fine Crystalline Powder |
198.0°C to 201.0°C | |
Authentic | |
CH3C6H4CH=CHCO2H | |
09, 617 | |
RURHILYUWQEGOS-VOTSOKGWSA-N | |
(E)-3-(4-methylphenyl)prop-2-enoic acid | |
731767 | |
99% | |
4-Methylcinnamic acid, Predominantly trans |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 217-479-9
- 4-Methylcinnamic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.