Learn More
4-Methylcyclohexanone, 98%, Thermo Scientific Chemicals
£16.80 - £40.40
Chemical Identifiers
CAS | 589-92-4 |
---|---|
Molecular Formula | C7H12O |
Molecular Weight (g/mol) | 112.17 |
MDL Number | MFCD00001643 |
InChI Key | VGVHNLRUAMRIEW-UHFFFAOYSA-N |
Synonym | 4-methylcyclohexanone, cyclohexanone, 4-methyl, 4-methyl-1-cyclohexanone, methyl-4 cyclohexanone-1, p-methyl cyclohexanone, methycyclohexanone, metylocykloheksanon, unii-0l1r78k79s, 4-methyl cyclohexanone, methyl-4 cyclohexanone-1 french |
PubChem CID | 11525 |
IUPAC Name | 4-methylcyclohexan-1-one |
SMILES | CC1CCC(=O)CC1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
589-92-4 | |
112.17 | |
VGVHNLRUAMRIEW-UHFFFAOYSA-N | |
11525 | |
CC1CCC(=O)CC1 |
C7H12O | |
MFCD00001643 | |
4-methylcyclohexanone, cyclohexanone, 4-methyl, 4-methyl-1-cyclohexanone, methyl-4 cyclohexanone-1, p-methyl cyclohexanone, methycyclohexanone, metylocykloheksanon, unii-0l1r78k79s, 4-methyl cyclohexanone, methyl-4 cyclohexanone-1 french | |
4-methylcyclohexan-1-one |
Specifications
589-92-4 | |
100.0 | |
Colorless to Yellow | |
169°C to 171°C | |
Authentic | |
Glass bottle | |
CH3C6H9(=O) | |
MFCD00001643 | |
0.914 | |
VGVHNLRUAMRIEW-UHFFFAOYSA-N | |
4-methylcyclohexan-1-one | |
11525 | |
98% | |
4-Methylcyclohexanone, 98% |
97.5 | |
-41°C | |
0.9140g/mL | |
40°C | |
98% | |
C7H12O | |
1.4445 to 1.4465 | |
07,18 | |
4-methylcyclohexanone, cyclohexanone, 4-methyl, 4-methyl-1-cyclohexanone, methyl-4 cyclohexanone-1, p-methyl cyclohexanone, methycyclohexanone, metylocykloheksanon, unii-0l1r78k79s, 4-methyl cyclohexanone, methyl-4 cyclohexanone-1 french | |
CC1CCC(=O)CC1 | |
112.17 | |
112.17 | |
Liquid |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 209-665-3
missing translation for 'rtecsNumber' : GW1750200
missing translation for 'tsca' : TSCA
- 4-Methylcyclohexanone
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Flammable liquid Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only