Learn More
4-Methylmorpholine N-oxide, 50 wt.% solution in water, Thermo Scientific Chemicals
£16.30 - £118.00
Specifications
Chemical Name or Material | 4-Methylmorpholine N-oxide |
---|---|
Name Note | 50 wt.% solution in water |
CAS | 7529-22-8, 7732-18-5 |
CAS Min % | 50.0, 48.0 |
CAS Max % | 52.0, 50.0 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10823201
|
Thermo Scientific Acros
348270250 |
25 g | Glass bottle |
£16.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10745062
|
Thermo Scientific Acros
348271000 |
100 g | Glass bottle |
£22.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10735062
|
Thermo Scientific Acros
348270010 |
1 kg | Glass bottle |
£118.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentSpezifikation
4-Methylmorpholine N-oxide | |
7529-22-8, 7732-18-5 | |
52.0, 50.0 | |
Colorless to Yellow | |
118.0°C to 119.0°C | |
Glass bottle | |
MFCD00005947 | |
15, 6171 | |
4-methylmorpholine n-oxide, 4-methylmorpholine 4-oxide, n-methylmorpholine oxide, n-methylmorpholine n-oxide, morpholine, 4-methyl-, 4-oxide, 4-methylmorpholine-4-oxide, methyl morpholine oxide, n-methylmorpholine-n-oxide, nmo, unii-arc64pkj0f | |
LFTLOKWAGJYHHR-UHFFFAOYSA-N | |
4-methyl-4-oxidomorpholin-4-ium | |
82029 | |
117.15 |
50 wt.% solution in water | |
50.0, 48.0 | |
-20.0°C | |
1.1300g/mL | |
>100°C | |
C5H11NO2 | |
07,244 | |
1.13 | |
Solubility in water: miscible | |
C[N+]1([O-])CCOCC1 | |
117.15 | |
CHEBI:52093 | |
Liquid |
Sicherheit und Handhabung
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 231-391-8
- 4-Methylmorpholine N-oxide, 50% wt.% solution in water
Signalwort
- Warning
Gefahrenkategorie
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Gefahrenhinweis
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Sicherheitshinweis
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only