Learn More
4-Methylpiperidine, 98+%, Thermo Scientific Chemicals
£29.04 - £100.10
Chemical Identifiers
CAS | 626-58-4 |
---|---|
Molecular Formula | C6H13N |
Molecular Weight (g/mol) | 99.18 |
MDL Number | MFCD00006005 |
InChI Key | UZOFELREXGAFOI-UHFFFAOYSA-N |
Synonym | 4-pipecoline, piperidine, 4-methyl, gamma-pipecoline, 4-methylpiperidin, .gamma.-pipecoline, 4-methyl-piperidine, 4-methylpiperdine, 4-methylpyperidine, 4-methyl piperidine, hexahydro-g-picolineShow More |
PubChem CID | 69381 |
IUPAC Name | 4-methylpiperidine |
SMILES | CC1CCNCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11307805
|
Thermo Scientific Alfa Aesar
L02709.AE |
100 mL |
£29.04
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11317805
|
Thermo Scientific Alfa Aesar
L02709.AP |
500 mL |
£100.10
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Methylpiperidine acts as a catalyst in the preparation of stilbene derivatives. It is used as an anti-corrosive compound for iron. It is also used in the synthesis of peptides to remove or deprotection of 9-fluorenylmethyloxycarbonyl (Fmoc) groups.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methylpiperidine acts as a catalyst in the preparation of stilbene derivatives. It is used as an anti-corrosive compound for iron. It is also used in the synthesis of peptides to remove or deprotection of 9-fluorenylmethyloxycarbonyl (Fmoc) groups.
Solubility
Miscible with water and chloroform.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, carbon dioxide and strong oxidizing agents.
Chemical Identifiers
626-58-4 | |
99.18 | |
UZOFELREXGAFOI-UHFFFAOYSA-N | |
69381 | |
CC1CCNCC1 |
C6H13N | |
MFCD00006005 | |
4-pipecoline, piperidine, 4-methyl, gamma-pipecoline, 4-methylpiperidin, .gamma.-pipecoline, 4-methyl-piperidine, 4-methylpiperdine, 4-methylpyperidine, 4-methyl piperidine, hexahydro-g-picolineShow More | |
4-methylpiperidine |
Specifications
626-58-4 | |
0.838 | |
123°C to 124°C | |
Amine-like | |
1.4458 | |
UN2733 | |
4-pipecoline, piperidine, 4-methyl, gamma-pipecoline, 4-methylpiperidin, .gamma.-pipecoline, 4-methyl-piperidine, 4-methylpiperdine, 4-methylpyperidine, 4-methyl piperidine, hexahydro-g-picoline | |
UZOFELREXGAFOI-UHFFFAOYSA-N | |
4-methylpiperidine | |
69381 | |
≥98% |
4°C to 5°C | |
13 | |
13°C (55°F) | |
C6H13N | |
MFCD00006005 | |
1083 | |
Miscible with water and chloroform. | |
CC1CCNCC1 | |
99.18 | |
99.18 | |
4-Methylpiperidine |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302-H311+H331-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 210-954-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.