missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Methylsulfonyl)aniline, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
500 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used as pharmaceutical intermediate. A series of 4-(methylsulfonyl)aniline derivatives were synthesized in order to obtain new compounds as a potential anti-inflammatory agents with expected selectivity against COX-2 enzyme.
Solubility
Soluble in water.
Notes
Light sensitive. Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
It is used as pharmaceutical intermediate. A series of 4-(methylsulfonyl)aniline derivatives were synthesized in order to obtain new compounds as a potential anti-inflammatory agents with expected selectivity against COX-2 enzyme.
Solubility
Soluble in water.
Notes
Light sensitive. Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications
Melting Point | 133°C to 138°C |
Color | White |
Density | 1.44 g/mL |
Quantity | 25 g |
UN Number | UN2811 |
Beilstein | 2208425 |
Sensitivity | Light sensitive |
Solubility Information | Soluble in water. |
IUPAC Name | 4-methanesulfonylaniline |
Formula Weight | 171.21 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'