missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonylamino)benzeneboronic acid, 95%, Thermo Scientific™

Product Code. 15419797
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 380430-57-9
Molecular Formula C7H10BNO4S
Molecular Weight (g/mol) 215.03
MDL Number MFCD02179473
InChI Key NDVJJEADFLTFCD-UHFFFAOYSA-N
Synonym 4-methylsulfonylamino phenylboronic acid, 4-methylsulfonamido phenylboronic acid, 4-methanesulfonylaminophenylboronic acid, 4-methylsulfonylaminophenylboronic acid, 4-methanesulfonylamino phenylboronic acid, 4-methylsulfonamido phenyl boronic acid, 4-methanesulfonamidophenyl boronic acid, 4-methanesulfonamido phenyl boronic acid, 4-methylsulphonyl amino benzeneboronic acid, 4-methylsulfonylamino benzeneboronic acid
PubChem CID 2773537
IUPAC Name [4-(methanesulfonamido)phenyl]boronic acid
SMILES CS(=O)(=O)NC1=CC=C(C=C1)B(O)O

Specifications

Melting Point 148°C to 154°C
Quantity 1 g
Formula Weight 215.03
Percent Purity 95%
Chemical Name or Material 4-(Methylsulfonylamino)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'