missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)-m-cresol, 97%, Thermo Scientific Chemicals

£17.59 - £61.90

Chemical Identifiers

CAS 3120-74-9
Molecular Formula C8H10OS
Molecular Weight (g/mol) 154.227
MDL Number MFCD00045773
InChI Key VKALYYFVKBXHTF-UHFFFAOYSA-N
Synonym 3-methyl-4-methylthio phenol, 4-methylthio-m-cresol, methylthiomethylphenol, mmtp, phenol, 3-methyl-4-methylthio, 3-methyl-4-1-methylthio phenol, usaf ma-17, 4-methylthio-3-methylphenol, m-cresol, 4-methylthio, 3-methyl-4-methylsulfanyl phenol
PubChem CID 18391
ChEBI CHEBI:38681
IUPAC Name 3-methyl-4-methylsulfanylphenol
SMILES CC1=C(C=CC(=C1)O)SC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11301877
View Documents
Thermo Scientific Alfa Aesar
L07353.06
5 g
£17.59
5g
Please to purchase this item. Need a web account? Register with us today!
11311877
View Documents
Thermo Scientific Alfa Aesar
L07353.14
25 g
£61.90
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3120-74-9
154.227
VKALYYFVKBXHTF-UHFFFAOYSA-N
18391
3-methyl-4-methylsulfanylphenol
C8H10OS
MFCD00045773
3-methyl-4-methylthio phenol, 4-methylthio-m-cresol, methylthiomethylphenol, mmtp, phenol, 3-methyl-4-methylthio, 3-methyl-4-1-methylthio phenol, usaf ma-17, 4-methylthio-3-methylphenol, m-cresol, 4-methylthio, 3-methyl-4-methylsulfanyl phenol
CHEBI:38681
CC1=C(C=CC(=C1)O)SC

Specifications

3120-74-9
C8H10OS
1210322
VKALYYFVKBXHTF-UHFFFAOYSA-N
3-methyl-4-methylsulfanylphenol
18391
154.23
4-(Methylthio)-m-cresol
54°C to 58°C
MFCD00045773
3-methyl-4-methylthio phenol, 4-methylthio-m-cresol, methylthiomethylphenol, mmtp, phenol, 3-methyl-4-methylthio, 3-methyl-4-1-methylthio phenol, usaf ma-17, 4-methylthio-3-methylphenol, m-cresol, 4-methylthio, 3-methyl-4-methylsulfanyl phenol
CC1=C(C=CC(=C1)O)SC
154.227
CHEBI:38681
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P312-P332+P313-P361-P363-P501c

H311-H315-H319

missing translation for 'einecsNumber' : 221-496-7

missing translation for 'rtecsNumber' : GP2225000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only