missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl-alpha-D-galactopyranoside hydrate, 99%

Product Code. 10162921 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 mg
250 mg
Packaging:
Glass bottle
Unit Size:
250mg
50mg
This item is not returnable. View return policy

Product Code. 10162921

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 38597-12-5 | C16H18O8 | 338.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38597-12-5
Molecular Formula C16H18O8
Molecular Weight (g/mol) 338.31
MDL Number MFCD03791284
InChI Key YUDPTGPSBJVHCN-HWSQZMMLNA-N
Synonym 4-methylumbelliferyl-alpha-d-galactopyranoside, 4-methyl-7-2r,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy-2h-chromen-2-one, 4-methyl-7-2r,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy chromen-2-one, 7-alpha-d-galactopyranosyloxy-4-methyl-2h-1-benzopyran-2-one, 4-methylumbelliferyl a-d-galactopyranoside, 4-methylumbelliferyl alpha-d-galactoside, 4-methylumbelliferyl-a-d-galactopyranoside, 4-methylumbelliferyl alpha-d-galactopyranoside, 4-methyl-2-oxo-2h-1-benzopyran-7-yl alpha-d-galactopyranoside, 4-methylumbelliferyl alpha-d-galactopyranoside tlc
PubChem CID 2733788
IUPAC Name 4-methyl-7-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one
SMILES CC1=CC(=O)OC2=CC(O[C@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)=CC=C12

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 50 mg
Formula Weight 338.31
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylumbelliferyl-α-D-galactopyranoside hydrate, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.